Learn More
MilliporeSigma Milli-Q™ Polishing Kit for Elix™ Pure Water
Description
- This purification kit contains IPAK Meta™ and IPAK Quanta™ polishing cartridges for Elix™ water, designed to work as a pair to produce ultrapure (Type 1) water for sensitive laboratory applications.
- The cartridges contain a combination of Jetpore™ mixed-bed ion-exchange resin and innovative IQnano™ ion-exchange media to remove trace ions from purified water, as well as a high-grade synthetic activated carbon, which targets trace organic contaminants.
Specifications
Specifications
| Feedwater Source | Elix Purified Water |
| Purification Method | Ultrapure |
| Resin Type | Ion Exchange Resin |
| Type | Polishing Kit |