missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Milli-Q™ IX Pure Water System with E-POD™ Dispenser

The most advanced pure water system for the production of Elix™ quality water, E-POD™ pure water dispenser included.

Supplier:  MilliporeSigma™ ZIX7005P0C

 View more versions of this product

Catalog No. ZIX7005P0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Applications

  • Milli-Q™ IX Water Purification Systems produce pure (Type 2) water for critical lab applications
  • Ideal for microbiology, histology, cytology, immunohistochemistry (IHC), electrophoresis gel and buffers, western blotting, immunoassays (ELISA, RIA), drug dissolution testing, biological oxygen demand (BOD), chemical oxygen demand (COD), and UV/Vis spectroscopy
  • Use to prepare samples for laser fluorometry, in the synthesis and characterization of membranes for fuel cells, or in the synthesis of nanocomposites by electrochemistry
  • For general lab applications including sample preparation, buffer and reagent preparation, glassware rinsing, etc.
  • Feed laboratory instruments such as autoclaves, dishwashers, weathering and stability test chambers, clinical analyzers and slide stainers, hydrogen generators, and ultrapure water systems (ex. Milli-Q™ IQ 7000 System)

Features and Benefits

  • A compact, ergonomic, mercury-free, and intelligent water purification solution that delivers consistently high quality pure water on demand
  • Pure water production unit offers flexible installation and simplified maintenance
  • Pure water storage tank allows maximum protection from any external source of contamination
  • Multi-targeted protection from bacterial, particulate, and CO2 contamination by using a bactericidal UV lamp, vent filter, automatic sanitization module (ASM), and an overflow sensor
  • Large touch screen allows for intuitive interaction and displays essential data for complete user reassurance
  • Requires a specific country box with the user manual in local language and a power cord adapted to the local electrical network (Sold separately)

E-POD™ Dispenser

  • Provides pressurized pure water, up to 2 L/min
  • 3 different final filters can be connected to target specific contaminants and remove them right when dispensing pure water
  • 5 in. touchscreen with bright, colored LCD display is easy to read and convenient to use, even with gloves on
  • Navigation in operating and dispensing modes is intuitive, without spending time sorting through multiples menus to find information
  • Graphic icons on the display allow all users to quickly and easily operate the system
  • Comprehensive information displayed on the screen ensures water is always dispensed at the desired quality
  • Variable flow provides 2 different flows: low flow and full flow up to 2 L/min
  • Volumetric water dispensing automatically delivers a specific water volume, from 20 mL to 100 L (in 1 mL increments)
  • Up to two E-POD™ dispensers can be used with each IX production unit at different locations within the same laboratory, or in adjacent laboratories

Note:
A complete water purification system is comprised of multiple components including the purification unit, consumables, and in some cases, dispensers, tanks, and other accessories, which should be included in your sales quotation. Please contact your Fisher Scientific Representative or MilliporeSigma Lab Water Solutions Specialist to receive a comprehensive quote prior to ordering.

Specifications

Chemical Identifiers

<2000 μS/cm at 25°C
1/2 in. Gaz
Potable Tap
1 to 6 bar
IEC13 Female
41°F to 95°F
100/240 V
<0.001 EU/mL
≤10 CFU/L
Pretreated Water Using Electrodeionization (EDI), Deionization (DI), Reverse osmosis (RO), or Distillation Technologies
≤30 ppb
4 to 10
350 V (Max)
5°C to 35°C
50/60 Hz
Water Purification System

Specifications

<2000 μS/cm at 25°C
1/2 in. Gaz
Potable Tap
≤30 ppb
Milli-Q™ IX 7005
IEC13 Female
41°F to 95°F
770 x 342 x 520 mm (30.3 x 13.4 x 20.5 in.)
50/60 Hz
Water Purification System
≤10 CFU/L
Pretreated Water Using Electrodeionization (EDI), Deionization (DI), Reverse osmosis (RO), or Distillation Technologies
5 L/hr.
1 to 6 bar
4 to 10
350 V (Max)
5°C to 35°C
100/240 V
<0.001 EU/mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions