Learn More
MilliporeSigma™ Milli-Q™ IX 7003 Water Purification System
The most advanced pure water system producing Elix™ water quality at 3 L/h to be connected to Milli-Q™ IQ Storage Tank and E-POD™ remote dispenser to deliver pure (Type 2) water at up to 2 L/min. Intelligent, intuitive and green (RO/EDI/UV mercury free).
Supplier: MilliporeSigma™ ZIX7003T0C
Description
The Milli-Q™ IX 7003 has been designed in a compact, ergonomic, mercury-free and intelligent way to deliver high quality pure water.
The system features three separate and distinct components:
- Milli-Q™ production unit offers flexible installation and a more simplified maintenance.
- The Milli-Q™ IQ Storage Tank and Top Tank is the optimal solution to protect pure water.
- The new E-POD™ point-of-delivery unit provides pure (Type 2) water through its large, intuitive touch screen. Up to two E-POD™ dispensers can be with each production unit at different locations in the laboratory.
Pure (Type 2) water product allows to perform many critical and general lab applications:
- Microbiology culture media preparation, staining solutions for histology and cytology, immunohistochemistry (IHC), electrophoresis gel and buffers, western blotting, immunoassays (ELISA, RIA), drug dissolution testing, biological oxygen demand (BOD), chemical oxygen demand (COD), UV/Vis spectroscopy, titration, etc.
- Sample preparation (dilution, extraction...), buffer and reagent preparation, glassware rinsing, etc.
- Feeding laboratory instruments like autoclaves, dishwashers, weathering & stability test chambers, clinical analyzers and slide stainers, hydrogen generators, and ultrapure water systems (e.g. Milli-Q™ IQ 7000 system)
DIGITAL SERVICES SIMPLIFY SYSTEM CARE AND ASSURE PRODUCTIVITY
MyMilli-Q™ Remote Care is a unique online monitoring and service feature that provides:
- Remote access to your system and its data
- Automatic notifications, by email or SMS, in case of an alert or alarm
- Faster service with remote diagnostics and repair possible
- Simplified data management and advanced traceability that eases audit and accreditation processes
Note: A complete water purification system is comprised of multiple components including the purification unit, consumables, and in some cases, dispensers, tanks, and other accessories, which should be included in your sales quotation. Please contact your Fisher Scientific Representative or MilliporeSigma Lab Water Solutions Specialist to receive a comprehensive quote prior to ordering.
Specifications
| ≤10 CFU/L | |
| Potable Tap | |
| Potable Tap | |
| <30 ppb | |
| 3 L/hr. (Milli-Q™ IX 7003), 5 L/hr. (Milli-Q™ IX 7005), 10 L/hr. (Milli-Q™ IX 7010), 15 L/hr. (Milli-Q™ IX 7015) |
| Up to 2 L/min. from POD | |
| 3 L/hr. (Milli-Q™ IX 7003), 5 L/hr. (Milli-Q™ IX 7005), 10 L/hr. (Milli-Q™ IX 7010), 15 L/hr. (Milli-Q™ IX 7015) | |
| >5 Mohm.cm | |
| <30 ppb |