Learn More
MilliporeSigma™ Milli-Q™ IX 7003/5/10/15 US Country Box
Catalog No. ZFCBIXXUS
Change view
Click to view available options
For Use With (Equipment):
Milli-Q IX 7003/05/10/15 Water Purification System
Catalog No. For Use With (Equipment)
ZFCBIXXUS Milli-Q IX 7003/05/10/15 Water Purification System
1 options
Catalog No. ZFCBIXXUS Supplier MilliporeSigma™ Supplier No. ZFCBIXXUS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Water Purification Consumables Kit, compatible with the Milli-Q™ IX 7003 and Milli-Q™ IX 7005 systems.
Water Purification Consumables Kit, compatible with the Milli-Q™ IX 7003 and Milli-Q™ IX 7005 systems and includes:
  • IPAK GARD™ H 3-5 Pretreatment Pack (hard water)
  • Activated Carbon for protection against chlorine, organics, bacteria
  • Polyphosphate for protection against scaling and precipitates
  • Depth Filter for protection against particulates and colloids
  • Tank Vent Filter
  • Activated Carbon for volatile organics removal
  • Soda Lime for CO2 removal
  • Membrane Filter to stop particulates and bacteria
Specifications
For Use With (Equipment) Milli-Q IX 7003/05/10/15 Water Purification System
Type Country Box Milli-Q™ IX 7003/5/10/15 US
Includes A Quick Reference Guide and a user manual in local language on a USB key, and a power cord adapted to the local electrical network