Learn More
MilliporeSigma™ Milli-Q™ IQ 7000 Qualification Workbook
IQ/OQ Qualification Package for Milli-Q IQ 7000 system
Supplier: MilliporeSigma™ VIQ7000FQ
Description
Milli-Q™ IQ 7000 Purification System. An ultrapure water solution designed with YOU in mind.
Simple and intuitive dispensing
- Just a twist of the new dispensing wheel lets you deliver water at the flow rate you need, from precise drop by drop up to a full 2 liters per minute. For added efficiency, you can also directly select a dispensing option from the Home screen.
- While dispensing, you can easily check the water quality parameters, always visible onscreen. Their values are continuously monitored by the system, so you can have full confidence in your water quality.
Complementary technologies
- The Milli-Q™ IQ 7000 system combines advanced technologies, hydraulic design and software capabilities to efficiently produce the superior-quality ultrapure water you need to work confidently.
A powerful combination of purification media
- The slim IPAK Meta™ and IPAK Quanta™ cartridges are designed to function as a pair. The cartridges use a combination of Jetpore™ mixed-bed ion-exchange resin and innovative IQnano™ ion-exchange media to provide exceptional performance.
Water quality to match all your application needs
- You'll find that our range of POD-Paks is the perfect solution for your various applications. Each of the Application-Pak polishers targets specific types of contaminants and removes them right at the Q-POD™ dispenser.
Versatile configurations for the best fit
- Each lab is unique. The Milli-Q™ IQ 7000 system lets you optimize valuable lab space to help maximize your efficiency.
Easy data management for a paperless environment
- Powerful, integrated data management makes it so much simpler and faster to manage your water system data – no more log book or paper to archive! All data generated by the Milli-Q™ IQ 7000 is stored in the system memory.
Order Info
Must be ordered with the Milli-Q™ IQ 7000 Ultrapure Water System. Please contact your Fisher Rep for more details.
- Environmental benefits include:
- Renewable Energy
Specifications
IQ/OQ Qualification Workbook |