Learn More
MilliporeSigma™ Milli-Q™ EQ Wall Mounting Kit
Save precious laboratory space by wall-mounting the accessories for your EQ 7000 Water Purification System.
$406.00 - $2,200.00
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
|
WMEQ0DKT
|
MilliporeSigma™
WMEQ0DKT |
Milli-Q™ EQ 7000 Touchscreen |
Each for $406.00
|
|
|||||
|
WMEQ0RKT
|
MilliporeSigma™
WMEQ0RKT |
Milli-Q™ EQ 7000 Q-POD Dispenser |
Each for $2,200.00
|
|
|||||
Description
The Q-POD™ Dispenser or HMI Touchscreen for the Milli-Q™ EQ 7000 Water Purification System can be wall-mounted up to 3 m from the system, saving precious space.
- Includes all necessary components for convenient installation of the Q-POD™ Dispenser or HMI Touchscreen for the Milli-Q™ EQ 7000 Water Purification System
- Optimizes installation flexibility of the system, allowing setup to adapt to ever-changing laboratories
Specifications
| Wall Mounting Kit |