missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Miller™ Revolution™ Harnesses

Catalog No. p-3004428
Change view
Click to view available options
Buckle Style (Leg):
Quick-Connect Buckle Leg Straps
Tongue-Buckle Leg Straps
Material:
DualTech Webbing
Dupont™ Kevlar™ aramid fiber-Nomex™ aramid fibers webbing
Style:
Black with Dupont™ Kevlar™-Nomex™ aramid fibers webbing
Standard Harness
With Belt, Side D-rings and Pad
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Buckle Style (Leg) Material Style
191304080 Quick-Connect Buckle Leg Straps DualTech Webbing Standard Harness
191304086 Quick-Connect Buckle Leg Straps DualTech Webbing With Belt, Side D-rings and Pad
191304081 Tongue-Buckle Leg Straps DualTech Webbing Standard Harness
191304104 Quick-Connect Buckle Leg Straps Dupont™ Kevlar™ aramid fiber-Nomex™ aramid fibers webbing Black with Dupont™ Kevlar™-Nomex™ aramid fibers webbing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 191304080 Supplier Honeywell™ Supplier No. RDTQC/UBKU
Only null left

Design based on key user-identified needs

  • PivotLink™ connection with unique rotary design promotes greater comfort in bending/mobility; also serves as an attachment point for belts, tools and accessories as part of an integrated accessory system
  • Cam buckle provides easy, one-handed adjustment/release and allows simultaneous adjustment of shoulder straps
  • Semi-flexible ErgoArmor™ back shield minimizes impact from sharp/heavy snap hooks and self-retracting lifelines; knitted mesh membrane minimizes moisture absorption and maintains optimal air circulation
  • Stand-up back D-ring simplifies connection
  • DualTech™ webbing makes donning easy with two-sided, contrasting-color webbing and textures
  • High-impact-resistant polymer web finials provide positive gripping surface for easy web adjustment
  • Integrated self-contained label pack encapsulates labels, decreasing damage and loss

Order Info

Webbing tested to 7000 lb. tensile strength. Universal size.

Compliance

Meets OSHA 1910.66 and OSHA 1926.502 (d); ANSI™ A10.32 and ANSI Z359-2007; CSA Z259.10 requirements.

Specifications

Color Black
Material DualTech Webbing
Product Type Revolution Harness
Style Standard Harness
For Use With (Application) Construction, Fall Protection
Size Universal
Buckle Style (Leg) Quick-Connect Buckle Leg Straps