missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Miller™ Revolution™ DualTech™ Harnesses

Catalog No. 19165283
Change view
Click to view available options
D-Ring Location:
Front
Front, Side
Includes:
Integrated accessory system
Pad, integrated accessory system
Removable belt and pad, integrated accessory system
Size:
2X-Large/3X-Large
Universal
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. D-Ring Location Includes Size
19165283 Front Integrated accessory system Universal
19165289 Front, Side Pad, integrated accessory system Universal
19165292 Front, Side Removable belt and pad, integrated accessory system Universal
19165294 Front, Side Removable belt and pad, integrated accessory system 2X-Large/3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19165283 Supplier Honeywell™ Supplier No. RDTFD TB/UBKU

Harnesses with DualTech webbing offering safety and functionality with features developed to meet key user needs

  • Available with tongue buckle legs and front and/or side D-rings
  • Available with or without removable belt
  • Exceed worker expectations in comfort, fit, ease-of-use, style, durability, compliance, flexibility and convenience

Specifications

Color Black
Size Universal
D-Ring Location Front
Includes Integrated accessory system
Style Full-body harness
Webbing Material DualTech™, Polyester
For Use With (Application) Construction, manufacturing and transportation fall protection
Buckle Style (Leg) Tongue-Buckle Leg Straps
Certifications/Compliance OSHA, ANSI and CSA