Learn More
Honeywell™ Miller™ Revolution™ Arc-Rated Harnesses
Catalog No. 19169921
Change view
Click to view available options
Product Type:
Arc-Rated Harness without Rescue Loops
Size:
Universal
Catalog No. Product Type Size
19169921 Arc-Rated Harness without Rescue Loops Universal
1 options
Catalog No. 19169921 Supplier Honeywell™ Supplier No. RKNAR QC/UBK
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Protect from falls caused by electric arc flash and arc blast exposure
  • Kevlar™/Nomex™ DualTech™ webbing has 10,000 lb. tensile strength; engineered with permanent fire-retardant properties
  • Kevlar outer offers durability and fire resistance while Nomex inner gives greater comfort
  • PivotLink™ design allows freedom of motion while cam buckles provide easy, two-finger adjustment
  • Feature quick-connect chest and leg buckles, cam buckles, a self-contained label pack, web finials, a Kevlar web back D-ring, and an ErgoArmor™ back shield
  • Leather insulators under all-metal hardware increase worker protection
  • Bright red binding on every pad for easy identification
Order Info
Universal size. Harnesses are rated to 400 lb. capacity.
Certifications
Meet ASTM™ F887-05 arc-flash standard.
Compliance
Meets OSHA 1910.66 and OSHA 1926.502 (d); ANSI™ A10.32 and ANSI Z359-2007; ASTM™ F887-13; CSA Z259.10 requirements.
Specifications
Material Dupont™ Kevlar™ aramid fiber, Nomex™ aramid fibers
Certifications/Compliance Meets ASTM F887-05 arc-flash standard; Meet OSHA and CSA requirements.
For Use With (Application) Designed to protect workers from falls caused by electric arc-flash and arc-blast exposure
Size Universal
Product Type Arc-Rated Harness without Rescue Loops