missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Miller™ Polyester Body Belts

Harness body belts featuring a tongue buckle with 11 grommets

$116.81 - $116.81


Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-034811
View Documents
Honeywell™
6414NUBK
Universal
Each for $116.81
Only null left
 
Description

Description

  • Size: Large or Universal
  • Color: Black
  • Material: 44mm (1.75 in.) nylon
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'