Learn More
Honeywell Safety Products™ Miller™ HP™ Fall Arrest and Retrieval Harnesses
Catalog No. 19274601
Change view
Click to view available options
Buckle Style:
Qwik-Fit
D-Ring Location:
Back
Size:
Large/X-Large
Catalog No. Buckle Style D-Ring Location Size
19274601 Qwik-Fit Back Large/X-Large
1 options
Catalog No. 19274601 Supplier Honeywell Safety Products™ Supplier No. 850T 4UGKC
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Webbing coated with PTFE to extend service life
Superior water and oil repellency and protection against dirt, grime, spills and stains.
  • With lanyard ring, sub-pelvic strap, back strap and back or side D-rings
  • Bright green for high visibility and ease of inspection
  • Friction buckles on the shoulder straps
(This style is also available with front D-ring, side D-rings and mating buckles on the shoulder straps. See below.) Models 850T and 850T-7 have mating buckles on both the leg and chest straps. Models 850T-4 and 850T-58 have tongue buckles on the leg straps and mating buckles on the chest strap.
Compliance
Meet all applicable OSHA, ANSI, and CSA requirements.
Specifications
Material PTFE-coated Nylon Webbing
Certifications/Compliance Meets all applicable OSHA, ANSI, and CSA requirements.
Size Large/X-Large
Buckle Style Qwik-Fit
D-Ring Location Back