Learn More
Honeywell™ Miller™ Bos'n Chair
Manual Descent Device is designed for a variety of applications where smooth, controlled descent from an elevated position is required. Features 3-inch wide webbing and 3/4-inch thick plywood seat and measures 12 x 24 in.
Supplier:  Honeywell™ 6004NSYL
Catalog No. 19020560
Only null left
Explore available promotions
Description
Description
  • Controlled by the user, allowing for fast or slow decent
  • Decelerate and stop by applying slight outward pressure on the free end of the line
  • Lock in a stationary position for a hands-free work environment
  • Device must be ordered with Miller brand descent rope
  • Meets all applicable OSHA, ANSI and CSA standards
Specifications
Specifications
Bosn's chair
OSHA, ANSI, CSA
Product Suggestions
Product Suggestions