missing translation for 'onlineSavingsMsg'
Learn More

Miglitol 98.0+%, TCI America™

Catalog No. p-7133146
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23021G 1 g
M23025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23021G Supplier TCI America Supplier No. M23021G
Only null left

Chemical Identifiers

CAS 72432-03-2
Molecular Formula C8H17NO5
Molecular Weight (g/mol) 207.23
MDL Number MFCD00867240
InChI Key IBAQFPQHRJAVAV-ULAWRXDQSA-N
PubChem CID 441314
ChEBI CHEBI:6935
IUPAC Name (2R,3R,4R,5S)-1-(2-hydroxyethyl)-2-(hydroxymethyl)piperidine-3,4,5-triol
SMILES OCCN1C[C@H](O)[C@@H](O)[C@H](O)[C@H]1CO

Specifications

Melting Point 146°C
Color White-Yellow
Quantity 1 g
Formula Weight 207.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Miglitol
RTECSNumber TN4350170
TSCA No