Learn More
Fisherbrand™ Micryo™ Labels Strips and Dots for Cryo Storage, Laser Printer Sheets
GREENER_CHOICE
Intended uses: cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications.
 View more versions of this product
Catalog No. 15930E
Only null left
Includes: Template for printing instructions.
Description
Description
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications.
Ideal for all cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications. Suitable for use on microcentrifuge tubes, cryovials, racks, boxes and microtiter plates. Adhesive withstands temperatures from -196° to 121°C (-321° to 250°F)
  • No need to self-laminate
  • Thinner labels allow tubes to fit in most storage systems
  • Available in a full range of colors and sizes
  • Guaranteed not to fall off under normal use (provided user instructions are followed)
  • Also offered in rolls (see Cat. Nos. 15-910 and 15-920 Series)
Cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications
Specifications
Chemical Identifiers
Cryogenic Label
Cryostorage, Liquid Nitrogen, refrigeration and ultra-low temperature freezer applications
-321 to 250°F
-196°C to +121°C
Specifications
White
Cryogenic Label
Racks, boxes
1 x 2.6 in.
-321 to 250°F
1 in. x 2.6 in.
Label
Cryostorage, Liquid Nitrogen, refrigeration and ultra-low temperature freezer applications
600/Pk.
2.5 x 6.6 cm
-196°C to +121°C
Product Suggestions
Product Suggestions