missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Microvolume Pipet Tips, Sterile

Catalog No. 07200524
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.5 to 10 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
07200524 0.5 to 10 μL
07200521 0.1 to 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07200524 Supplier Corning™ Supplier No. 4830

Provide accurate, reliable performance in the stated volume range

  • Microvolume tips provide accurate, reliable performance in the 0.1 μL to 10 μL range for major brand ultra-micropipettors.
  • RNase-/DNase-free.
  • Nonpyrogenic.

Specifications

Volume (Metric) 0.5 to 10 μL
Filtered Non-filtered
Sterility Sterile
Color Natural
Autoclavable Autoclavable
Certifications/Compliance EN ISO 13485 compliant, FDA CFR 21 Part 820 compliant (cGMP), BSE/TSE compliant, Non-Pyrogenic, USP Class VI tested, DNase/RNase Free, Quality Control Tested
Clean Claims DNase, RNase, and Pyrogen free
Format Rack
Tip Style Microvolume, Round
Packaging Rack
Quantity 96/Pk., 960/Cs.
Product Type Pipette Tips
For Use With (Equipment) Eppendorf, Universal Fit Ultramicro
Material Polypropylene
Show More Show Less