Learn More
Research Products Corporation Disposable Plastic Transfer Pipettes, Extended Fine Tip, 1.5ml, 400 per Package
Catalog No. 50550200
Encompass
Plastic transfer pipettes are molded into one-piece units from low density polyethylene. They eliminate cross-contamination encountered with rubber bulbs and reduce exposure when working with hazardous or infectious samples. Ideal for all laboratory procedures that require fast and safe transferring of liquids. Pipettes can also be used as freeze vials or as storage and transport containers by merely heat sealing the tips closed after filling. They can be sterilized if required by gas or radiation. Pipettes are supplied non-sterile. Specially designed to fit micro-tubes. Micro-design allows tip to reach bottom of tube, assay plates and electrophoresis wells. Eliminates air bubbles when filling.
Catalog No. 50550200 Supplier Research Products Corporation Supplier No. RPI 147500
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Plastic transfer pipettes are molded into one-piece units from low density polyethylene. They eliminate cross-contamination encountered with rubber bulbs and reduce exposure when working with hazardous or infectious samples. Ideal for all laboratory procedures that require fast and safe transferring of liquids. Pipettes can also be used as freeze vials or as storage and transport containers by merely heat sealing the tips closed after filling. They can be sterilized if required by gas or radiation. Pipettes are supplied non-sterile. Specially designed to fit micro-tubes. Micro-design allows tip to reach bottom of tube, assay plates and electrophoresis wells. Eliminates air bubbles when filling.