missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MicroTest™ M4RT 3 mL w/o Beads Tube, Red

Catalog No. R12700
Change view
Click to view available options
Quantity:
72 Tubes/Pk
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R12700 72 Tubes/Pk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R12700 Supplier Thermo Scientific™ Supplier No. R12700

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Maintain viability of viruses chlamydiae, mycoplasma and urealasma with top quality transport and storage media that comes in four versatile formulations

Maintain organism viability and counts for verification studies, quality control procedures, and repeat testing of viruses, chlamydiae, mycoplasma and ureaplasma with top quality media from the experts in media. Thermo Scientific™ MicroTest™ Tubes come in four media formulations to offer flexibility in both transport and storage.

Designed for the collection, transportation, maintenance and long-term storage of viral specimens, chlamydiae, mycoplasma and ureaplasma, Thermo Scientific MicroTest products are top quality. From formulation to packaging, MicroTest products maintain the viability of organisms through freeze-thaw cycles while inhibiting antimicrobial contaminants and promoting safety.

MicroTest™ M4RT™ contains gelatin, gentamicin, and amphotericin B for the transport of viruses and Chlamydiae.

MicroTest Tubes come with 1.5 mL or 3 mL of liquid medium in a 15 mL conical tube with 3 glass beads.

Keep your lab moving in the right direction with a dependable set of solutions for transport, holding, maintenance and long-term culture storage with the MicroTest family of collection and transport media.

Trusted reliable results

  • Stringent quality control, including cellular toxicity on each lot number
  • Patented formulations with stabilizing proteins, buffers, and specific antibiotics

Compatibility and viability

  • Provides compatible transport for cell culture, enzyme immunoassays, and molecular diagnostic tests
  • Maintains organism viability and counts for better, more reliable results during validation/verification studies, quality control procedures, and repeat testing

Convenient, accommodating format

  • 1.5 mL and 3.0 mL fill volumes allow multiple tests from one specimen
  • Individual transport media tubes or complete kits offer flexibility and versatility

Specifications

Closure Color Red
Compatibility Culture, Shell vial, Freeze-thaw cycles, Culture maintenance, ELISA, PCR, DNA probe technology
Description M4RT 3 mL Tube without Beads, Red
For Use With (Application) Chlamydiae Viruses
Packaging Packaged in partitioned boxes
Quantity 72 Tubes/Pk
Storage Requirements 2°C to 30°C
Transport Temperature 2° to 30°C
Volume (Metric) 3 mL
Green Features ACT Ecolabel (Gold)