Learn More
Qsonica Microson XL2000 Microprobes
Supplier:  Qsonica P 3
 View more versions of this product
Catalog No. 15338268
Only null left
Description
Description
For use with Misonix™ Microson™ XL2000 Ultrasonic Homogenizers.
Specifications
Chemical Identifiers
Microprobe
Misonix™ Microson™ XL-2000 ultrasonic homogenizer
Specifications
Microprobe
0.2 to 5mL
0.24 cm
14.3 cm
210 μm
0.09 in.
5.6 in.
Misonix™ Microson™ XL-2000 ultrasonic homogenizer
Product Suggestions
Product Suggestions