Learn More
Supertek™ Microslide™ - Nerve and Muscle Action Set
Description
- Image titles on Microslide:
- 1. Single (100x)
- 2. Synapse (left side - 100x) (right side - 3,000,000x)
- 3. Axons (left side - x.s. - 400x) (right side - I.s. -200x)
- 4. Myelinated Axons (600x)
- 5. Nerve-Muscle Junction (400x)
- 6. Sunmaptic Knobs - E.M. (300,000x)
- 7. Striated Muscle Cell (1000x)
- 8. Sarcomeres (left side relaxed - E.M. 45,000x) (right side - contracted - E.M. 31,000x)
- Lesson Plan with Pocket to store Microslide
- Reproducible Worksheet with a purchase of 15
Specifications
Specifications
| Type | Nerve Muscle Action Microslide |
| Surface Treatment | Untreated |
| Material | Paper and Film |
| Dimensions (LxW) | Lesson Plan 6.5 x 8 in., Slide 2 x 8 in. |
| Product Line | Supertek Microslides |
| Sterility | Non-sterile |