missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ MicroSEQ™ ID Software for SeqStudio™ Genetic Analyzer v1.0
Description
- Allows users to easily identify and classify unidentified bacterial or fungal sequences by comparing them to a validated microbial library
- Automatically identifies unknown specimens
- Features a validated library of over 11,000 microbial sequences (with latest libraries)
- MicroSEQ™ ID sequence libraries consist of bacterial libraries of 500-bp 16S rDNA sequences and over 9500 validated entries
- Extensive coverage of gram-negative non-fermenters, Bacillus, coryneforms, mycobacteria, and Staphylococcus
- Libraries contain only correctly identified organisms and quality-controlled sequences derived from type strains of preeminent culture collections
- Supports sequence polymorphisms and includes bacterial Category A biothreat agents
- Run a plate on the SeqStudio™ Genetic Analyzer directly via the MicroSEQ™ ID Software
- MicroSEQ™ system includes security, audit, and e-signature features
- Qualification and validation support available
Specifications
Specifications
| Product Line | MicroSEQ ID Software |
| Description | Includes digital download letter and license code kit |
| For Use With (Application) | Microbial risk mitigation, identification |
| For Use With (Equipment) | QST SeqStudio™ Instrument with SAE |
| Libraries | Not included |
| License | Included |
| Quantity | 1 each |
Frequently Asked Questions (FAQs)
Does MicroSEQ ID Microbial Identification Software work for RPO beta regions?
No, MicroSeq ID Software does not work for RPO beta regions.
Does MicrobeBridge Software (Cat. No. A29238) require MicroSEQ ID Microbial Identification Software to be installed in order to function?
No, MicrobeBridge Software does not require MicroSeq ID Software to also be installed.
Which Thermo Fisher Scientific instrument laptops are compatible with MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 (Lite Installation)?
The Dell Latitude 3541 Laptop with WIN10 IOT 2018-64-bit is the Thermo Fisher Scientific instrument laptop that is compatible with MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 (Lite Installation).
Which Thermo Fisher Scientific instrument computers are compatible with MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 (System and Lite Installations)?
The Dell Optiplex XE3 with WIN10 IOT 2018-64-bit is the Thermo Fisher Scientific instrument computer that is compatible with MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 (System and Lite Installations).
Which KB Basecaller version is supported for MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0?
KB Basecaller v1.4.2.5 is supported for MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0