Learn More
Applied Biosystems™ MicroSEQ™ ID Sequencing Clean-up Plates
Description
The MicroSEQ™ ID Sequencing Clean-up Plates are used for post-sequencing clean-up in the MicroSEQ™ ID workflow. The gel filtration matrix used in the microplate cartridges provides a convenient and simple way to remove dye terminators, dNTPs, and salts, as well as residual primers from cycle sequencing reactions.
The MicroSEQ™ ID Sequencing Clean-up Plates can be used downstream of the following sequencing kits:
- MicroSEQ™ 500 16S rDNA Sequencing Kit
- MicroSEQ™ Full Gene 16S rDNA Sequencing Kit
- MicroSEQ™ D2 rDNA Fungal Sequencing Kit
This product provides two 96-well microplate cartridges ideal for high throughput microbial identification workflows. The end products can be directly used for sequencing on any Applied Biosystems™ genetic analyzers.
Specifications
Specifications
| Quantity | 2 x 96-well plates |
| For Use With (Equipment) | 3500xL Genetic Analyzer, MicroSEQ™ ID, 3500 Genetic Analyzer |
| Product Type | Microplate |
| Shipping Condition | Wet Ice |
| Content And Storage | Store in refrigerator (2°C to 8°C). |
For Research Use Only. Not for use in diagnostic procedures.