This microscope slide storage cabinet is constructed of heavy gauge steel with a baked-on beige enamel finish. Cabinet is stackable using interlocking tabs and can be stacked compatibly with similar cabinets already in use. Each cabinet has six individual drawers with a total capacity of up to 5,000 (3 x 1 Inches) glass slides.
The drawers have black phenolic pull knobs. Cabinet construction prevents drawers from falling out accidentally, but they can be disengaged for use at a work area. Foam rubber dividers keep slides upright when drawers are partially filled. Identification tabs are included for labeling. Each slide cabinet is fully inspected before shipment and is packaged using recyclable, biodegradable materials.