Learn More
AirClean™ Systems Microscope Enclosures, Upflow
Catalog No. 361018793
Change view
Click to view available options
Dimensions (D x W x H):
60.9 x 121.9 x 106.7 cm
60.9 x 81.3 x 101.6 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H)
361018793 60.9 x 81.3 x 101.6 cm
361018905 60.9 x 121.9 x 106.7 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361018793 Supplier AirClean™ Systems Supplier No. AC632TMIC
Only null left
The upflow microscope enclosure protects the operator from gases, fumes or particulates with bonded carbon or HEPA filtration.
  • Designed to work with range of microscopes
  • Microscope access port allows for easy access of the eyepiece and viewing platform
  • Optional sturdy mobile cart
  • Microprocessor controlled
  • Audible and visible alarms
  • Constructed from UV and chemical resistant material
  • Custom sash closure allows for maximum operator protection without compromising airflow
  • Workstations are delivered fully assembled ready for use, simply plug in and use
  • Process protection workstations equipped with safety interlock
  • Available in downflow and upflow versions
Specifications
Product Type Microscope Enclosure
Air Supply Upflow
Dimensions (D x W x H) 60.9 x 81.3 x 101.6 cm
For Use With (Application) Protects operator from exposure to harmful particulate
Includes Microprocessor controlled; Audible and visible alarms; UV/chemical resistant; Safety interlock