missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Microscope C-Mount Adapter

Compound C-Mount Adapter
$76.65 - $178.84
Specifications
| Type | C-mount Adapter |
|---|---|
| Warranty | 5 years optical and mechanical, 1 year electrical from date of purchase |
| Catalog Number | Magnification Power | Size | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Magnification Power | Size | Price | Quantity & Availability | |||||
03000056
![]() |
0.3X | C-Mount 23 mm, 1/3 in. Chip |
Each for $76.65
|
|
|||||
03000102
![]() |
0.3X | 1/3 in. Chip |
Each for $153.31
|
|
|||||
03000062
![]() |
0.5X | C-Mount 23 mm, 1/2 in. Chip |
Each for $153.31
|
|
|||||
|
03000063
|
0.3X | C-Mount 21 mm, 1/3 in. Chip |
Each for $127.78
|
|
|||||
03000065
![]() |
0.5X | C-Mount 21 mm, 1/2 in. Chip |
Each for $178.84
|
|
|||||
Description
- Accessory for microscope
Warranty
1 Year
Specifications
| C-mount Adapter | |
| 5 years optical and mechanical, 1 year electrical from date of purchase |