missing translation for 'onlineSavingsMsg'
Learn More

Surgical Design™ Microprobe

Catalog No. 22079740
Change view
Click to view available options
Tip Style:
Needle-Sharp tips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
22079740 Needle-Sharp tips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22079740 Supplier Surgical Design™ Supplier No. 28308P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Surgical Design™ Microprobe Set is for micromanipulation in the lab.

  • Each microprobe is precision-ground stainless steel that has been tempered for quality control and experimental requirements
  • Tips are needle-sharp and have been hand-honed
  • Set includes six different microprobes

Specifications

Product Type Micro Probe
Autoclavable Autoclavable
Color Silver
Material Stainless Steel
For Use With (Application) Micro tissue manipulation
Quantity Set of 6
Tip Style Needle-Sharp tips