Learn More
MicroGuard MP™ Microporous Coveralls with Attached Hood and Boots
Catalog No. 19472780
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Catalog No. Size
19472780 Small
19472779 Medium
19472778 Large
19472781 X-Large
19472774 2X-Large
19472775 3X-Large
19472776 4X-Large
19472777 5X-Large
8 options
Catalog No. 19472780
Supplier: MicroGuard MP™ 8019S
Only null left
Microporous coveralls with attached hood and boots deliver protection and comfort through a strong and durable fabric.
  • Comfortable state-of-the-art lamination technology allows for maximum breathability.
  • Soft non-woven fabric combined with a microporous film allows moisture vapor to escape, so heat and moisture are not trapped near the body.
  • Attached hood and boots, along with elastic wrists and ankles, provide extra protection of the neck, shoulders, and feet.
  • Coveralls offer base-level protection against light liquid splashes and nonhazardous particulates providing the optimal balance of comfort, durability, and protection.
  • Roomy, seamless shoulder area resists tears and provides more protection in key exposure areas.
  • Wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric.
  • Elastic back stretches with user for more comfort and better tear resistance.
  • Elastic wrists and ankles offer a secure fit to help prevent skin exposure.
  • Environmentally friendly, can be recycled after use.
Specifications
Product Type General Purpose Coveralls
Size Small
Gender Unisex
Material Microporous Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Protection Class General
Resistance Abrasion
Model 8019
No. of Pockets None
Show More Show Less