Learn More
MicroGuard MP™ Microporous Coveralls with Attached Hood
Microporous coveralls with attached hood deliver protection and comfort through a strong and durable fabric.
$178.47 - $227.85
Specifications
Product Type General Purpose Coveralls
Gender Unisex
Material Microporous Fabric
Color White
Disposable Single-use
View More Specs
Products 9
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19472772
View Documents
MicroGuard MP™
8015S
Small
Case of 25 Each for $178.47
Only null left
19472771
View Documents
MicroGuard MP™
8015M
Medium
Case of 25 Each for $178.47
Only null left
19472770
View Documents
MicroGuard MP™
8015L
Large
Case of 25 Each for $178.47
Only null left
19472773
View Documents
MicroGuard MP™
8015XL
X-Large
Case of 25 Each for $178.47
Only null left
19472765
View Documents
MicroGuard MP™
80152XL
2X-Large
Case of 25 Each for $182.53
Only null left
19472766
View Documents
MicroGuard MP™
80153XL
3X-Large
Case of 25 Each for $194.62
Only null left
19472767
View Documents
MicroGuard MP™
80154XL
4X-Large
Case of 25 Each for $204.13
Only null left
19472768
View Documents
MicroGuard MP™
80155XL
5X-Large
Case of 25 Each for $214.37
Only null left
19472769
View Documents
MicroGuard MP™
80156XL
6X-Large
Case of 25 Each for $227.85
Only null left
Description
Description
  • Comfortable state-of-the-art lamination technology allows for maximum breathability.
  • Soft non-woven fabric combined with a microporous film allows moisture vapor to escape, so heat and moisture are not trapped near the body.
  • Attached hood, along with elastic wrists and ankles, provide protection of the neck and shoulders and a secure fastening in high-movement areas where skin exposure can happen.
  • Coveralls offer base-level protection against light liquid splashes and nonhazardous particulates providing the optimal balance of comfort, durability, and protection.
  • Roomy, seamless shoulder area resists tears and provides more protection in key exposure areas.
  • Wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric.
  • Elastic back stretches with user for more comfort and better tear resistance.
  • Elastic wrists and ankles offer a secure fit to help prevent skin exposure.
  • Environmentally friendly, can be recycled after use.
  • Attached hood provides additional protection of the neck and shoulders.
Specifications
Specifications
General Purpose Coveralls
Microporous Fabric
Single-use
Elastic
Hooded
Front Zipper with Storm Flap
Non-sterile
Abrasion
None
Unisex
White
Elastic
Elastic
Attached
Serged
General
8015
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'