missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Microplates for Fluorescence-based Assays

Catalog No. M33089
Encompass
Change view
Click to view available options
No. of Wells:
384
96
Quantity:
10 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells Quantity
M33089 96 10 Plates
M33091 384 10 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M33089 Supplier Invitrogen™ Supplier No. M33089

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Microplates for fluorescence-based assays are 96- and 384-well, black-walled, clear-bottom microplates that are optimized for fluorescence-based applications on standard fluorescence microplate readers.

Microplates for fluorescence-based assays are black-walled, clear-bottom microplates that minimize well-to-well crosstalk and autofluorescence. These 96- and 384-well microplates are compatible with standard fluorescence microplate readers and optimized for fluorescence-based applications including nucleic acid quantitation (e.g., Quant-iT assays, including PicoGreen, RiboGreen, and OliGreen).

Features of the microplates for fluorescence-based assays include:

  • Black-walled design—minimizes well-to-well crosstalk and autofluorescence
  • Clear-bottom wells—allows for optimal fluorescence detection
  • Compatibility—suitable for standard fluorescence microplate readers
  • Optimized for fluorescence microplate assays—ideal for nucleic acid quantitation (e.g., Quant-iT assays)
  • Versatile formats—available in 96- and 384-well configurations

Specifications

Barcode No Barcode
Format Standard
Content And Storage Store at room temperature.
Array 8 x 12
Autoclavable Not Autoclavable
Packaging 10-plates
Product Line Invitrogen
Color Clear Bottom, Black
Disposable Single-use
Product Type Microplates
For Use With (Equipment) Microplate Reader
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 200 μL
Sterility Non-sterile
Well Shape Flat Bottom (F-well)
Quantity 10 Plates
Show More Show Less
What kind of 96-well plate do I need for my fluorescent assay?

Select a microplate that is compatible with your instrument. Refer to the manufacturer's manual for types of microplates that are recommended for use with your instrument.

Various types of plastic may autofluoresce. Select a microplate that is rated for fluorescence-based assays and exhibits little or no autofluorescence in the emission range of the reagent you wish to use.

Clear plates are routinely used for fluorescence-based assays, but opaque-walled microplates reduce bleed-through between wells. If you are using live cells, you may need to consider special substrate treatments for the bottom of the wells, such as poly-L-lysine.

For Research Use Only. Not for use in diagnostic procedures.