missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Microplate Seals
Description
Self-sticking with inert adhesive. Whatman™ Microplate Seals prevent spillage and reduce sample evaporation.
- Controls humidity and reduces evaporation of samples.
- Prevents spills and contamination.
- Cold seals are self-sticking with inert adhesive.
- Heat seals are available in a clear polyester or aluminum foil.
- Heat seals are for polypropylene microplates only and are applied with heat and pressure.
Specifications
Specifications
| Color | Clear |
| Material | Polyester |
| No. per Pack | 100 Pieces |
| For Use With (Application) | Used to control humidity and reduce evaporation of samples |
| Product Type | Thin Cold Sealing Film with Adhesive Backing |
| Thickness | 0.05 mm |