Learn More
Thermo Scientific™ Sealing Tape for 96-Well Plates
Catalog No. PI15036
Change view
Click to view available options
Quantity:
100 sheets
Catalog No. Quantity
PI15036 100 sheets
1 options
Catalog No. PI15036 Supplier Thermo Scientific™ Supplier No. 15036
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Clear, self-sticking sealing tape for standard microplates.
Clear, self-sticking sealing tape for standard microplates.
Thermo Scientific Microplate Sealing Tape is a package of pre-cut, pressure-sensitive sheets for sealing the surface of standard 96-well or 384-well microplates during incubation steps.
Features of Microplate Sealing Tape:
• Pressure-sensitive—position the tape over plate and press to seal; easy to remove
• Separate sheets—each sheet of tape is supplied on its own easy-to-peal backing (liner)
• Durable and inert—film thickness provides support and has a thin layer of acrylic adhesive
Use the disposable sheets to seal and reseal a microplate two or three times for incubation steps in an ELISA or protein assay procedure. Seal plates quickly and easily to allow mixing or prevent evaporation during assay incubation steps.
Specifications
Color Clear
Material Polyester
Sterility Non-sterile
No. of Wells 96/384
Quantity 100 sheets
Product Type Film, Seal, or Applicator
Adhesive Pressure Sensitive
Plate Compatibility 96/384-well Plate
For Research Use Only. Not for use in diagnostic procedures.