Learn More
Thermo Scientific™ Aluminum Support, tube/plate rack, 384-well
Thermo Scientific™ aluminum support rack can be used for sample preparation, heat-sealing and automatic dispensing.
Supplier: Thermo Scientific™ PRK1383*
Description
Perform sample preparation, heat-sealing and automatic dispensing using Thermo Scientific™ aluminum support rack.
Available in 96- and 384-well format.
- For heat-sealing and automated dispensing
- Industry-standard footprint fits in robotic workstations
- Can be heated or cooled to assist with sample preparation
Includes:
Recommended for:
Sample handling, sample preparation, heat-sealing, automated dispensing
Specifications
| Microplate Rack | |
| Microplates (384-well) | |
| Rectangular | |
| 1 rack |
| Blue/Clear, Clear | |
| Aluminum | |
| Real-Time PCR Systems or Thermal Cyclers |