missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Microplate Rotor for Thermo Scientific Sorvall™ Legend™ Mach 1.6™/1.6R Centrifuges

Catalog No. 75002010
Change view
Click to view available options
Max. Speed:
4,400 rpm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Max. Speed
75002010 4,400 rpm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 75002010 Supplier Thermo Scientific™ Supplier No. 75002010
Only null left

Includes

Rotor and lid

For use with Legend Mach 1.6 and 1.6R Centrifuges.

  • Capacity: Four microplates or two deepwell plates
  • Max. Speed/RCF: 4400rpm/3355 × g
  • Biocontainment-approved with optional lid 75-002-011 (Sold separately, in pairs)
  • Microplates sold separately

Specifications

Type MP 3300 Microplate Rotor
Max. RCF 3,355 x g
Max. Speed 4,400 rpm