Learn More
World Precision Pre-Pulled Glass Pipettes-TIP01TW1F
Catalog No. 50821944
Encompass
WPI can quickly supply your need for consistently sized, pre-pulled, glass, calibrated micropipettes. Tip diameters (ID) range from 0.1 to 10 m. Silanized Tips (Luer Shank) are available. Silanization waterproofs the glass to retard water when inserting into cell. This will not let the outside fluid run down the glass pipettes and get inside so easily.These glass needles are fabricated from Schott Duran borosilicate glass. 
Catalog No. 50821944 Supplier World Precision Supplier No. TIP01TW1F
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
WPI can quickly supply your need for consistently sized, pre-pulled, glass, calibrated micropipettes. Tip diameters (ID) range from 0.1 to 10 m. Silanized Tips (Luer Shank) are available. Silanization waterproofs the glass to retard water when inserting into cell. This will not let the outside fluid run down the glass pipettes and get inside so easily.These glass needles are fabricated from Schott Duran borosilicate glass.