Learn More
Micronova™ Scoops
Catalog No. 19165912
Change view
Click to view available options
Autoclavable:
Autoclavable
Capacity (Metric):
1100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable Capacity (Metric)
19165912 Autoclavable -
19165910 Autoclavable 1100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19165912 Supplier Micronova™ Supplier No. SCP02
Durable polypropylene scoop, available in sizes from 75 cc to 1100 cc. Ideal for scooping and sampling powders, granules, and pastes.
Micronova durable polypropylene scoops are available in sizes from 75 cc to 1100 cc and are ideal for scooping and sampling powders, granules, and pastes. Irradiated option available for sterile environments.
  • Individually bagged
  • Irradiated option available
Specifications
Product Type Scoop
Material Polypropylene
Autoclavable Autoclavable
Capacity (English) 6.72 oz.