missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Micrometer Screw Gauge

Catalog No. S24857
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S24857 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24857 Supplier Eisco™ Supplier No. PH0086B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Micrometer Screw Gauge with locking mechanism and ratchet top.

  • Nickel-plated brass
  • Accurately-machined stainless-steel rod
  • Supplied in case

Specifications

Grade 6 to Undergraduate
Includes Locking Mechanism
Material Nickel-plated Brass
Size 0 to 25 mm x 0.01 mm
Class Size 1 Lab Group
Product Type Micrometer Screw Gauge