missing translation for 'onlineSavingsMsg'
Learn More

MicroMax™ Microscopes

Catalog No. S90217A
Encompass
Change view
Click to view available options
Magnification Power:
60X to 100X
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
S90217A 60X to 100X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S90217A Supplier Carson Optical Supplier No. MM200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Requires

Two AAA batteries

Fit easily in your pocket to take anywhere.

Microscope has 60–100X magnification power with an extremely lightweight and portable design.

  • Built-in light provides a bright, clear image
  • Rubberized eyepiece for comfortable viewing
  • Weight: 1.28 oz. (36g)

Specifications

Lighting Type LED
Eyepiece Rubberized
Includes Built-in LED light, 3 SG3 button cell battery
Magnification Power 60X to 100X
Required Accessories Two AAA battery
Weight (English) 1.28 oz.
Weight (Metric) 36 g