Learn More
Epredia™ Microm™ Cryostats Heat Extractor
Catalog No. p-4529819
Change view
Click to view available options
For Use With (Equipment):
HM 525
HM 550 and 560
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
1407088 HM 550 and 560
361013375 HM 525
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1407088 Supplier Epredia™ Supplier No. 524510
Only null left
Use Epredia™ Microm™ Cryostats Heat Extractors with Epredia™ Cryostats.
  • Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen
  • Can be operated from any position on the freeze bar
  • Freezing time is shortened if freezing stage and tissue are placed on an active Peltier
Specifications
Description Heat Extractor for HM560
Product Type Heat Extractor
For Use With (Equipment) HM 550 and 560