missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One MICROLON™ 96-well ELISA Single-Break Strip Plates

Description
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Individual wells can be pipetted without leaving residue
- Flat bottom enables precise optical measurements
- Suitable for transmission measurements
Specifications
Specifications
| Barcode | Alphanumeric |
| Breakapart | Single break |
| Binding Property | High Binding |
| Format | Strip Plate |
| Array | 8 x 12 |
| Binding Type | High Binding |
| Color | Clear, Yellow |
| Product Type | Single-break Strip Plate |
| Material | Polystyrene |
| Volume (Metric) Well | 20 to 300 μL |
| Show More |