missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Microliter Rotor for Thermo Scientific Sorvall™ Legend™ Mach 1.6™/1.6R Centrifuges

Catalog No. 75003348
Change view
Click to view available options
Capacity:
48 x 1.5/2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
75003348 48 x 1.5/2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 75003348 Supplier Thermo Scientific™ Supplier No. 75003348

Includes

Rotor and lid

For use with Legend Mach 1.6 and 1.6R Centrifuges.

  • Capacity: 48 × 1.5/2mL
  • Max. Speed/RCF: 15,000rpm/24,652 × g
  • Biocontainment-approved
  • Adapters sold separately

Specifications

Type Microliter Rotor
Capacity 48 x 1.5/2 mL
Max. RCF 24,652 x g
Max. Speed 15,000 rpm
For Use With (Equipment) 1.6/1.6R Centrifuge