missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 9 mm Screw Thread Autosampler Vial Caps with FEP/Natural Rubber Septa

Catalog No. 15111032
Change view
Click to view available options
Color:
Black
Blue
Green
Natural
Red
Yellow
No. per Pack:
100
2000
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. per Pack
15111032 Yellow 2000
15111016 Blue 100
15111017 Blue 2000
15111019 Green 100
15111020 Green 2000
15111023 Black 2000
15111026 Natural 2000
15111028 Red 100
15111029 Red 2000
15111031 Yellow 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 15111032 Supplier DWK Life Sciences Supplier No. 090020YB

Available in multiple colors and package sizes.

Specifications

Color Yellow
Material Polypropylene
Pre-slit No
Septum FEP, Natural Rubber, FEP
No. per Pack 2000
Product Line MicroLiter
Closure Size 9 mm
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
Product Type Screw Cap