missing translation for 'onlineSavingsMsg'
Learn More

DWK LIFE SCIENCES™ MicroLiter 9 mm ScrewTin Autosampler Vial Caps with PTFE/Silicone Septa

Catalog No. 15111095
Change view
Click to view available options
Color:
Black
Blue
Green
Natural
Yellow
Pre-slit:
No
Yes
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Pre-slit
15111095 Natural Yes
15111074 Black No
15111068 Blue No
15111088 Blue Yes
15111071 Green No
15111077 Natural No
15111083 Yellow No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 15111095 Supplier DWK LIFE SCIENCES™ Supplier No. 090035NT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MicroLiter 9 mm ScrewTin Autosampler Vial Caps with PTFE/Silicone Septa are available in multiple colors and a pack size of 100. Designed for use with 9 mm screw-thread autosampler vials, these caps provide a reliable seal for chromatography applications.

  • ScrewTin caps designed for 9 mm screw-thread autosampler vials
  • PTFE/Silicone septa provide a reliable, low-extractable seal suitable for chromatography applications
  • Available in multiple cap colors: Blue, Green, Black, Natural, Red, and Yellow for easy sample identification and organization
  • Supplied in packs of 100
  • Pre-cut septa option available (09-0035NT) for easier needle penetration
  • Manufactured by DWK Life Sciences (WHEATON)

Specifications

Color Natural
Diameter (Metric) Outer 9 mm
Material Tinplate Steel
Pre-slit Yes
Sterility Non-sterile
Septum PTFE, Silicone
No. per Pack 100
Product Line MicroLiter
Quantity 100/Pk.
Closure Size 9 mm
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
Autoclavable Autoclavable
Product Type Screw Cap
Show More Show Less