missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 20 mm Pressure Release Crimp Top Headspace Vial Seals

Catalog No. 15111748
Change view
Click to view available options
Septum:
Aluminum, Silicone
Tan PTFE, White Silicone
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Septum
15111748 Aluminum, Silicone
15111737 Tan PTFE, White Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15111748 Supplier DWK Life Sciences Supplier No. 200060P

MicroLiter 20 mm crimp top seals are available in multiple colors and septa types.

Pressure release seal

Specifications

Color Silver
Material Aluminum
Pre-slit No
Septum Aluminum, Silicone
No. per Pack 100
Product Line MicroLiter
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Vials - MicroLiter
Product Type Crimp Cap