missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 16mm Septa for Screwthread Headspace Vial Caps

Catalog No. 15111605
Change view
Click to view available options
Color:
Blue
Gray
White
Septum:
PTFE
PTFE/Butyl
PTFE/Silicone
White PTFE/Tan Silicone
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Septum
15111605 White PTFE
15111606 Gray PTFE/Butyl
15111608 Blue PTFE/Silicone
15111611 Blue White PTFE/Tan Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 15111605 Supplier DWK Life Sciences Supplier No. 160010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MicroLiter septa for 16mm Screwthread Headspace Vials are constructed of PTFE, PTFE/butyl, or PTFE/silicone to suit a variety of chromatography applications.

Specifications

Color White
Pre-slit No
Septum PTFE
Quantity 100
Closure Size 16 mm
For Use With (Equipment) 16 mm Screw Thread Headspace Vials - MicroLiter