Learn More
International Enviroguard™ MicroGuard MP Microporous Shoe Cover with Non-skid Sole and Elastic Closure
Using state-of-the-art lamination technology, MicroGuard MP microporous shoe covers combine a soft, non-woven fabric with a microporous film to protect against spills and splashes.
Supplier: International Enviroguard™ 8105
Description
- Shoe covers are designed with a non-skid sole that helps reduce the risk of slips or falls
- A tear-resistant liquid and particulate barrier helps safeguard from blood penetration, various industrial chemicals, along with dust, dirt, and grime
- Breathable fabric allows moisture to escape, so your feet stay cool and comfortable even as you sweat
- Strong elastic closure on top helps keep these durable shoe covers in place, so your shoes and feet are protected throughout your workday
- Environmentally friendly; can be recycled after use
- Made of lightweight, tear-resistant polypropylene
- Passes ASTM F1670 testing for blood penetration
Specifications
| X-Large | |
| Yes | |
| Single-use | |
| Elastic Top | |
| Serged | |
| Shoe Covers |
| Multi-layer Barrier Laminate Fabric, Spunbond Polypropylene | |
| White | |
| Non-sterile | |
| Non-skid | |
| Static, Liquid, Grime, Dry Particulates |