Learn More
International Enviroguard™ MicroGuard MP Microporous Pants with Elastic Waist
Catalog No. 17184043
Change view
Click to view available options
Color:
White
Catalog No. Color
17184043 White
1 options
Catalog No. 17184043 Supplier International Enviroguard™ Supplier No. 82002XL
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
These disposable microporous pants are made with state-of-the-art lamination technology.
  • Soft, non-woven fabric is breathable, allowing moisture vapor and heat to escape so you stay cooler all shift long
  • Microporous pants provide a liquid and particulate barrier, offering high protection in non-hazardous work environments
  • Sport an elastic waist to keep pants secure as you work
  • No pockets
  • Environmentally friendly; can be recycled after use
  • Passes ASTM 1670 for blood penetration
  • Passes ASTM F903 for hydraulic jack oil, sodium hydroxide, mineral oil, bleach, isocyanate based paint hardener, and more
Specifications
Size 2X-Large
Gender Unisex
Product Type Chemical-resistant Pants
Material Multi-layer Barrier Laminate Fabric
Color White
Disposable Single-use
Waist Style Elastic