missing translation for 'onlineSavingsMsg'
Learn More

International Enviroguard™ MicroGuard MP Microporous Frock with Mandarin Collar, No Pockets

Catalog No. p-200019259
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17184074 Large
17184073 Medium
17184072 X-Large
17184078 2X-Large
17184077 3X-Large
17184076 4X-Large
17184075 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17184074 Supplier International Enviroguard™ Supplier No. 8048L
Only null left

Microporous Frock with elastic wrists and front snap closure offers the ideal balance of safety and comfort.

  • Fabric keeps dust and liquids out while allowing air and moisture to flow freely, so wearers stay cool throughout a hard day's work
  • Ideal for non-hazardous work environments that require reliable liquid and particulate protection
  • Frocks are produced with state-of-the-art lamination technology to combine a soft non-woven fabric with a microporous film that repels liquids
  • Frock does not have pockets
  • Secure, latex-free elastic wrists help keep sleeves in place to prevent skin exposure
  • Front snap closures are easy to secure
  • Low-linting fabric helps prevent cross-contamination
  • Possible applications include food processing, industrial applications, pharmaceuticals, cleanrooms and lab applications, and more

Specifications

Product Type Frock
Size Large
Material Microporous Fabric
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Snap (Front)
Cuff Style Elastic
Seam Type Serged
For Use With (Application) Keeping Out Dust and Liquids
Collar Type Mandarin
Latex Free Yes
Sleeve Style Long Sleeve
Autoclavable Not Autoclavable
Show More Show Less