Learn More
International Enviroguard™ MicroGuard MP 17 in. Microporous Boot Cover with Non-skid Sole and Elastic Closure
Using state-of-the-art lamination technology, MicroGuard MP microporous boot covers combine a soft, non-woven fabric with a microporous film that protects against splashes and spills.
$215.15 - $226.71
Specifications
Material Multi-layer Barrier Laminate Fabric, Spunbond Polypropylene
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-184-067
View Documents
International Enviroguard™
8104
Large
Case of 200 Each for $215.15
Only null left
17-184-065
View Documents
International Enviroguard™
8106
X-Large
Case of 200 Each for $226.71
Only null left
Description
Description
  • Boot covers are designed with a non-skid sole to help reduce the risk of slips or falls
  • A tear-resistant liquid and particulate barrier helps safeguard from blood penetration, various industrial chemicals, along with dust, dirt, and grime
  • Breathable fabric allows moisture to escape, so your feet stay more cool and comfortable even as you sweat
  • 17 in. length helps protect more of your lower legs from exposure to on-the-job contaminants
  • Secure elastic closure helps keep boot covers in place throughout your workday
  • Environmentally friendly; can be recycled after use
  • Strong, elastic top for a secure fit
  • Made of lightweight, tear-resistant polypropylene
  • Passes ASTM F1670 testing for blood penetration
Specifications
Specifications
Multi-layer Barrier Laminate Fabric, Spunbond Polypropylene
White
Non-sterile
Non-skid
Boot Covers
Yes
Single-use
Elastic Top with Ankle Ties
Serged
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'