Learn More
Contec™ MicroGenesis™ Mop Frame
GREENER_CHOICE
Catalog No. 19038566
Change view
Click to view available options
Dimensions:
10 x 44 cm (4 x 17.5 in.)
Catalog No. Dimensions
19038566 10 x 44 cm (4 x 17.5 in.)
1 options
Catalog No. 19038566 Supplier Contec™ Supplier No. MGMP0009
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
100% synthetic, disposable microfiber mop pad effectively removes fine particles and large debris from floors. Designed to be used wet or dry for general facilities cleaning in ISO Class 8 environments and above. Compatible with a wide range of disinfectants, including quats, peroxides and bleach.
  • Synthetic construction is compatible with common disinfectants such as quats, peroxides and bleach
  • New pad for each use simplifies quality control, inventory and space management, all without cross-contamination risk
  • Compatible with most existing hardware
Specifications
Dimensions 10 x 44 cm (4 x 17.5 in.)
Color Code White
Product Type Mop Frame
Color Blue
Sterility Non-sterile
Autoclavable Not Autoclavable
Disposable Single-use
Color-coded No
Material Polypropylene