missing translation for 'onlineSavingsMsg'
Learn More

Ansell MICROFLEX™ UltraSense™ EC USE-880 Nitrile Gloves

Extended cuff offers extra protection over the wrist and forearm for use in high-risk environments.

Supplier:  Ansell USE880XXL

 View more versions of this product

Catalog No. 19-167-295E

Only null left
Explore available promotions

Description

Description

High-quality nitrile offers reliable barrier protection against many hazards and infectious substances.

  • Patented ultra-soft nitrile provides the fit and feel of latex
  • Textured fingers ensure a secure grip
  • Extended, beaded cuff offers extra protection over wrist and forearm
  • Nonsterile, ambidextrous, single-use
  • Length: 11.4 in. (29 cm)
  • Fingertip thickness: 4.3 mil

Specifications

Specifications

ASTM D 6319
No
Blue
Partially Textured
0.13 mm
Nitrile
0.10 mm
2X-Large
0.13 mm
USE-880-XXL
33 MPa
29 cm
EN 420:2003 + A1:2009
Healthcare, Laboratory
Single-use
5.1 mil
Yes
3.9 mil
Powder Free
5.1 mil
Yes
UltraSense™ EC USE-880
11.4 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions