missing translation for 'onlineSavingsMsg'
Learn More

Ansell MICROFLEX™ UltraSense™ EC USE-880 Nitrile Gloves Edge
Edge

Extended cuff offers extra protection over the wrist and forearm for use in high-risk environments.

Supplier:  Ansell USE880M

 Afficher plus de versions de ce produit

Numéro de catalogue. 19-167-295B



Il en reste null
Edge
Découvrez plus d'offres spéciales

Description

Description

High-quality nitrile offers reliable barrier protection against many hazards and infectious substances.

  • Patented ultra-soft nitrile provides the fit and feel of latex
  • Textured fingers ensure a secure grip
  • Extended, beaded cuff offers extra protection over wrist and forearm
  • Nonsterile, ambidextrous, single-use
  • Length: 11.4 in. (29 cm)
  • Fingertip thickness: 4.3 mil

Spécifications

Spécifications

ASTM D 6319
No
Blue
Partially Textured
0.13 mm
Nitrile
0.10 mm
Medium
0.13 mm
USE-880-M
33 MPa
29 cm
EN 420:2003 + A1:2009
Healthcare, Laboratory
Single-use
5.1 mil
Yes
3.9 mil
Powder Free
5.1 mil
Yes
UltraSense™ EC USE-880
11.4 in.
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions